N-(2,2,2-Trifluoroethyl)-4-bromobenzylamine 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC50332-1G PC50332-250MG PC50332-5G
77430-908EA 454.4 USD
77430-908 77430-910 77430-912
N-(2,2,2-Trifluoroethyl)-4-bromobenzylamine 97%
N-(2,2,2-Trifluoroethyl)-4-bromobenzylamine
Formula: C₉H₉BrF₃N
Storage Temperature: Ambient
MDL Number: MFCD09928975
CAS Number: 728948-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR