4-Fluoro-2-cyclopropylphenol

Supplier: APOLLO SCIENTIFIC
Warning

PC32777-1G PC32777-250MG
77428-690EA 681.58 USD
77428-690 77428-692
4-Fluoro-2-cyclopropylphenol
4-Fluoro-2-cyclopropylphenol
Formula: C₉H₉FO
Storage Temperature: Freezer
MDL Number: MFCD24719781
CAS Number: 1232774-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR