1-(8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-3-yl)-2,2,2-trifluoroethanone

Supplier: APOLLO SCIENTIFIC
Warning

PC28278-1G PC28278-250MG PC28278-5G
77427-884EA 409.56 USD
77427-884 77427-886 77427-888
1-(8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-3-yl)-2,2,2-trifluoroethanone
1-(8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-3-yl)-2,2,2-trifluoroethanone
Formula: C₁₀H₃ClF₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD12406877
CAS Number: 1823182-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR