3-Fluoro-2-iodoanisole
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
2-Fluoro-6-methoxyiodobenzene, 3-Fluoro-2-iodophenyl methyl ether, 1-Fluoro-2-iodo-3-methoxybenzene
PC0873-1G
PC0873-25G
PC0873-5G
77412-708EA
38.26
USD
77412-708
77412-710
77412-712
3-Fluoro-2-iodoanisole
3-Fluoro-2-iodoanisole
Formula:
C₇H₆FIO Boiling Pt: 240 °C (746 mm Hg) Density: 1.859 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03788539 CAS Number: 7079-54-1 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...