But-2-ene-1,4-diamine, N-BOC protected 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (E)-tert-Butyl (4-aminobut-2-en-1-yl)carbamate

OR958835-1G OR958835-250MG
77412-194EA 155.88 USD
77412-194 77412-196
But-2-ene-1,4-diamine, N-BOC protected 95%
But-2-ene-1,4-diamine, N-BOC protected
Formula: C₉H₁₈N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09752966
CAS Number: 146394-99-2
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR