Chlorin e6

Supplier: APOLLO SCIENTIFIC
Warning

OR61545-100MG OR61545-250MG
77423-616EA 176.26 USD
77423-616 77423-618 77423-620
Chlorin e6
Chlorin e6
Formula: C₃₄H₃₆N₄O₆
MW: 596.673 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08669566
CAS Number: 19660-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR