N-(Toluene-4-sulfonyl)glycine methyl ester

Supplier: APOLLO SCIENTIFIC

Synonyms: Methyl N-[(4-methylphenyl)sulphonyl]glycinate, Tos-Gly-Ome, N-Tosylglycine methyl ester, Methyl [(4-methylphenyl)sulfonamido]acetate, Methyl [(4-methylphenyl)sulphonamido]acetate, Methyl N-[(4-methylphenyl)sulfonyl]glycinate

OR61063-100G OR61063-25G
77423-468EA 109.02 USD
77423-468 77423-470
N-(Toluene-4-sulfonyl)glycine methyl ester
N-(Toluene-4-sulfonyl)glycine methyl ester
Formula: C₁₀H₁₃NO₄S
Storage Temperature: Ambient
MDL Number: MFCD00558890
CAS Number: 2645-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR