2-Chloro-N,N-diisobutylacetamide

Supplier: APOLLO SCIENTIFIC
Warning

OR52317-10G OR52317-1G OR52317-5G
77422-648EA 174.22 USD
77422-648 77422-650 77422-652
2-Chloro-N,N-diisobutylacetamide
2-Chloro-N,N-diisobutylacetamide
Formula: C₁₀H₂₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD00040035
CAS Number: 5326-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR