3-(Piperidin-2-yl)aniline

Supplier: APOLLO SCIENTIFIC

OR321486-1G OR321486-250MG
77419-978EA 605.18 USD
77419-978 77419-980
3-(Piperidin-2-yl)aniline
3-(Piperidin-2-yl)aniline
Formula: C₁₁H₁₆N₂
Storage Temperature: Refrigerator
MDL Number: MFCD18702747
CAS Number: 1203797-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR