3-Amino-4-bromo-5-methylpyrazole 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 4-Bromo-5-methyl-1H-pyrazol-3-amine

OR943162-1G OR943162-25G OR943162-5G
77425-584EA 29.54 USD
77425-584 77425-586 77425-588
3-Amino-4-bromo-5-methylpyrazole 98%
3-Amino-4-bromo-5-methylpyrazole
Formula: C₄H₆BrN₃
MW: 176.02 g/mol
Melting Pt: 117…122 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03412169
CAS Number: 1780-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR