Hexafluoropropene trimer 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC4792-100G PC4792-25G
77415-716EA 212.94 USD
77415-716 77415-718
Hexafluoropropene trimer 97%
Hexafluoropropene trimer

Mixture of perfluoro-2,3,5-trimethylhexene-3,perfluoro-2,3,5-trimethylhexene-2 & perfluoro-2,4,5-trimethylhexene-2.

Formula: C₉F₁₈
Boiling Pt: 110…115 °C
Density: 1.83 g/cm³ (25 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00043821
CAS Number: 6792-31-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR