2,3,4,5-Tetramethyl-2-cyclopentenone 95%

Supplier: APOLLO SCIENTIFIC

OR3509-10G OR3509-25G OR3509-5G
77420-514EA 41.78 USD
77420-514 77420-516 77420-518
2,3,4,5-Tetramethyl-2-cyclopentenone 95%
2,3,4,5-Tetramethyl-2-cyclopentenone
Formula: C₉H₁₄O
Boiling Pt: 100 °C/30 mmHg
Density: 0.917 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00010248
CAS Number: 54458-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR