2-(4,5-Dichloro-1H-imidazol-1-yl)ethanethioamide

Supplier: APOLLO SCIENTIFIC
Warning

OR25115-1G OR25115-250MG
77407-588EA 135.5 USD
77407-588 77407-590
2-(4,5-Dichloro-1H-imidazol-1-yl)ethanethioamide
2-(4,5-Dichloro-1H-imidazol-1-yl)ethanethioamide
Formula: C₅H₅Cl₂N₃S
Storage Temperature: Ambient
MDL Number: MFCD00084996
CAS Number: 175201-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR