Furan-2-sulfonyl chloride 97+%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Furan-2-sulfonylchloride, Furan-2-sulphonylchloride

OR14486-250MG OR14486-1G
77404-974EA 144.68 USD
77404-974 77404-972
Furan-2-sulfonyl chloride 97+%
Furan-2-sulfonyl chloride
Formula: C₄H₃ClO₃S
Boiling Pt: 79…81 °C/10 mmHg
Storage Temperature: Freezer
MDL Number: MFCD06658968
CAS Number: 52665-48-2
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR