tert-Butyl-5-amino-1H-indazole-1-carboxylate
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
tert-Butyl 5-amino-1H-indazole-1-carboxylate
, N-(1)-Boc-5-aminoindazole
, 5-Amino-1H-indazole-1-carboxylic acid, 1,1 dimethyl ethyl ester
, 5-Amino-indazole-1-carboxylic acid tert-butyl ester
, 1-Boc-5-amino-indazole
OR30732-100MG
77419-102EA
13
USD
77419-102
tert-Butyl-5-amino-1H-indazole-1-carboxylate
tert-Butyl-5-amino-1H-indazole-1-carboxylate
Formula:
C₁₂H₁₅N₃O₂ MW: 233.27 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04114656 CAS Number: 129488-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...