2,2-Dimethyl-1-phenylpropylamine 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,2-Dimethyl-1-phenylpropan-1-amine

OR904924-1G OR904924-5G
77424-780EA 33.62 USD
77424-780 77424-782
2,2-Dimethyl-1-phenylpropylamine 98%
2,2-Dimethyl-1-phenylpropylamine
Formula: C₁₁H₁₇N
Storage Temperature: Ambient
MDL Number: MFCD00267378
CAS Number: 61501-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR