4-Chloro-5-methylpyrimidin-2-amine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR61403-1G OR61403-5G
77423-582EA 79.46 USD
77423-582 77423-584
4-Chloro-5-methylpyrimidin-2-amine 98%
4-Chloro-5-methylpyrimidin-2-amine
Formula: C₅H₆ClN₃
Melting Pt: 184…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00725898
CAS Number: 20090-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR