(m-Acrylamidophenyl)boronic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Acrylamidophenylboronic acid, (3-Acrylamidophenyl)boronic acid

OR40723-1G OR40723-5G
77421-200EA 37.7 USD
77421-200 77421-202
(m-Acrylamidophenyl)boronic acid 95%
(m-Acrylamidophenyl)boronic acid
Formula: C₉H₁₀BNO₃
MW: 191 g/mol
Melting Pt: 148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09025755
CAS Number: 99349-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR