N-[3-Aminomethyl)phenyl]acetamide hydrochloride 95+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-(Aminomethyl)acetanilide hydrochloride

OR7774-1G OR7774-250MG
77424-320EA 119.2 USD
77424-320 77424-322
N-[3-Aminomethyl)phenyl]acetamide hydrochloride 95+%
N-[3-Aminomethyl)phenyl]acetamide hydrochloride
Formula: C₉H₁₃ClN₂O
MW: 200.67 g/mol
Melting Pt: 230…233 °C
Storage Temperature: Ambient
MDL Number: MFCD04039171
CAS Number: 238428-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR