3-Chloro-2-(trifluoromethyl)-2-iodo-1,1,1-trifluoropropane

Supplier: APOLLO SCIENTIFIC
Warning

PC4948-1G PC4948-5G
77430-584EA 87.32 USD
77430-584 77430-586
3-Chloro-2-(trifluoromethyl)-2-iodo-1,1,1-trifluoropropane
3-Chloro-2-(trifluoromethyl)-2-iodo-1,1,1-trifluoropropane
Formula: C₄H₂ClF₆I
Boiling Pt: 124…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093304
CAS Number: 240122-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR