4-Fluoro-DL-phenylglycine 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Fluoro-DL-α-phenylglycine, 4-Fluorophenylglycine, H-DL-Phg(4-F)-OH

PC0378-5G PC0378-100G PC0378-25G
77426-236EA 21.36 USD
77426-236 77426-232 77426-234
4-Fluoro-DL-phenylglycine 98%
4-Fluoro-DL-phenylglycine
Formula: C₈H₈FNO₂
MW: 169.155 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00066444
CAS Number: 7292-73-1
EINECS: 230-724-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR