Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Fmoc-Lys-OH.HClNɑ-Fmoc-L-lysine HydrochlorideNα-Fmoc-L-lysine hydrochlorideFmoc-Lys-OH·HCl

OR925054-100G OR925054-25G OR925054-5G
77425-154EA 81.37 USD
77425-154 77425-152 77425-150
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride 98%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride
Formula: C₂₁H₂₄N₂O₄
MW: 404.89 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00190889
CAS Number: 139262-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR