Ferroceneboronic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR315060-1G OR315060-5G
77419-538EA 88.64 USD
77419-538 77419-540
Ferroceneboronic acid 95%
Ferroceneboronic acid
Formula: C₁₀H₁₁BFeO₂
MW: 229.85 g/mol
Melting Pt: 145…150 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00059074
CAS Number: 12152-94-2
EINECS: 235-272-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR