5,6-Dichloropyridine-3-sulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR4733-1G OR4733-250MG
77409-982EA 803.84 USD
77409-982 77409-984
5,6-Dichloropyridine-3-sulfonyl chloride
5,6-Dichloropyridine-3-sulfonyl chloride
Formula: C₅H₂Cl₃NO₂S
Melting Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD03541048
CAS Number: 98121-40-5
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR