(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochlorideH-Lys(Z)-OBzl·HClH-Lys(Z)-OBzl.HCl

OR350309-1G OR350309-25G OR350309-5G
77420-472EA 51 USD
77420-472 77420-474 77420-476
(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 97%
(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Formula: C₂₁H₂₆N₂O₄·HCl
MW: 406.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038981
CAS Number: 6366-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR