2-Propyl-1H-benzo[d]imidazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Propylbenzimidazole

OR110235-10G OR110235-5G
77403-158EA 225.16 USD
77403-158 77403-160
2-Propyl-1H-benzo[d]imidazole
2-Propyl-1H-benzo[d]imidazole
Formula: C₁₀H₁₂N₂
Melting Pt: 150…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00087477
CAS Number: 5465-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR