2-Imidazolecarboxylic acid 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR30610-25G OR30610-5G
77418-982EA 209.88 USD
77418-982 77418-984
2-Imidazolecarboxylic acid 96%
2-Imidazolecarboxylic acid
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Melting Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00270816
CAS Number: 16042-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR