Calcium pantothenate 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Calcium D-Pantothenate, D-Pantothenic acid calcium salt, D-Calcium pantothenate, Calcium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate, Vitamin B5 calcium salt, Calcium D-(+)-pantothenate

OR923775-100G OR923775-25G OR923775-5G
77425-102EA 54 USD
77425-102 77425-104 77425-106
Calcium pantothenate 95%
Calcium pantothenate
Formula: C₁₈H₃₂CaN₂O₁₀
MW: 476.54 g/mol
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00002766
CAS Number: 137-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR