4-Chloro-2,6-difluorobenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC8238-1G PC8238-5G
77433-616EA 21.42 USD
77433-616 77433-620
4-Chloro-2,6-difluorobenzaldehyde 98%
4-Chloro-2,6-difluorobenzaldehyde
Formula: C₇H₃ClF₂O
Melting Pt: 68…74 °C
Storage Temperature: Ambient
MDL Number: MFCD03094460
CAS Number: 252004-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR