4-Oxazolecarboxylic acid 98+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Oxazole-4-carboxylic acid

OR6277-5G OR6277-1G
77423-728EA 51 USD
77423-728 77423-726
4-Oxazolecarboxylic acid 98+%
4-Oxazolecarboxylic acid
Formula: C₄H₃NO₃
MW: 113.0715 g/mol
Melting Pt: 130…135 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD06797151
CAS Number: 23012-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR