8-Bromo-6-methylimidazo[1,2-a]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR6100-1G OR6100-5G
77423-440EA 66.22 USD
77423-440 77423-442
8-Bromo-6-methylimidazo[1,2-a]pyridine
8-Bromo-6-methylimidazo[1,2-a]pyridine
Formula: C₈H₇BrN₂
Melting Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD09258770
CAS Number: 136117-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR