2-Amino-6-methoxy-3-nitropyridine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 6-Methoxy-3-nitropyridin-2-amine

OR5827-100G OR5827-25G OR5827-5G
77423-228EA 137.54 USD
77423-228 77423-230 77423-232
2-Amino-6-methoxy-3-nitropyridine
2-Amino-6-methoxy-3-nitropyridine
Formula: C₆H₇N₃O₃
MW: 169.14 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD03206481
CAS Number: 73896-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR