4'-Benzyloxy-3'-fluoroacetophenone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(4-(Benzyloxy)-3-fluorophenyl)ethanone

PC10559-1G PC10559-5G PC10559-250MG
77426-706EA 252 USD
77426-706 77426-702 77426-704
4'-Benzyloxy-3'-fluoroacetophenone
4'-Benzyloxy-3'-fluoroacetophenone
Formula: C₁₅H₁₃FO₂
MW: 244.261 g/mol
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD06659550
CAS Number: 81227-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR