4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine

OR52321-5G OR52321-25G OR52321-1G
77422-660EA 47.38 USD
77422-660 77422-662 77422-664
4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine 98%
4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine
Formula: C₅H₄N₅I
MW: 261.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03787931
CAS Number: 151266-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR