6-Bromo-2-naphthylamine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Bromonaphthalen-2-amine, 2-Amino-6-bromonaphthalene

OR8620-1G OR8620-25G OR8620-5G
77424-578EA 99.84 USD
77424-578 77424-580 77424-582
6-Bromo-2-naphthylamine
6-Bromo-2-naphthylamine
Formula: C₁₀H₈BrN
MW: 222.0812 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01026466
CAS Number: 7499-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR