1-Bromo-2,4-bis(trifluoromethyl)benzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,4-Bis(trifluoromethyl)-1-bromobenzene

PC3105W-5G PC3105W-100G PC3105W-25G
77428-516EA 21.32 USD
77428-516 77428-512 77428-514
1-Bromo-2,4-bis(trifluoromethyl)benzene 97%
1-Bromo-2,4-bis(trifluoromethyl)benzene
Formula: C₈H₃BrF₆
MW: 293.01 g/mol
Boiling Pt: 158 °C/740 mmHg
Density: 1.738 g/cm³ (25 °C)
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00074904
CAS Number: 327-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR