1,1,2,3,3,3-Hexafluoropropyl dichloromethyl ether 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC9252-25G PC9252-5G
77434-262EA 135.5 USD
77434-262 77434-264
1,1,2,3,3,3-Hexafluoropropyl dichloromethyl ether 98%
1,1,2,3,3,3-Hexafluoropropyl dichloromethyl ether
Formula: C₄H₂Cl₂F₆O
Boiling Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD03094547
CAS Number: 56860-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR