3-Bromo-2-methylbenzotrifluoride 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Bromo-2-methyl-3-(trifluoromethyl)benzene

PC0486-1G PC0486-25G PC0486-5G
77426-248EA 21.26 USD
77426-248 77426-250 77426-252
3-Bromo-2-methylbenzotrifluoride 97%
3-Bromo-2-methylbenzotrifluoride
Formula: C₈H₆BrF₃
Boiling Pt: 35 °C/1 mmHg
Storage Temperature: Ambient
MDL Number: MFCD04038231
CAS Number: 69902-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR