Ethyl glyoxylate 50% in toluene, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

OR59968-500G OR59968-100G OR59968-25G
77410-654EA 115.36 USD
77410-654 77410-652 77410-650
Ethyl glyoxylate 50% in toluene, Technical Grade
Ethyl glyoxylate
Formula: C₄H₆O₃
Boiling Pt: 110 °C
Density: 1.003 g/cm³ (25 °C)
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00044009
UN: 1294
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR