Pyridoxal-5'-phosphate monohydrate 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: Pyridoxal-5-phosphate hydrate, Pyridoxal 5-phosphate monohydrate, Pyridoxal 5'-phosphate monohydrate, (4-Formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate hydrate

OR350314-100G OR350314-25G OR350314-5G
77420-484EA 289.34 USD
77420-484 77420-486 77420-488
Pyridoxal-5'-phosphate monohydrate 98%
Pyridoxal 5'-phosphate monohydrate
Formula: C₈H₁₀NO₆P·1H₂O
Storage Temperature: Freezer
MDL Number: MFCD00149414
CAS Number: 41468-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR