3-Iodooxetane

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Iodo-1,3-epoxypropane, 1,3-Epoxy-2-iodopropane

OR308004-1G OR308004-5G
77408-968EA 20.32 USD
77408-968 77408-970
3-Iodooxetane
3-Iodooxetane
Formula: C₃H₅IO
Boiling Pt: 166 °C
Density: 2.14 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD08544403
CAS Number: 26272-85-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR