3-(Trimethoxysilyl)propyl methacrylate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR323284-1KG OR323284-250G OR323284-500G
77420-124EA 96.78 USD
77420-124 77420-126 77420-128
3-(Trimethoxysilyl)propyl methacrylate 98%
3-(Trimethoxysilyl)propyl methacrylate
Formula: C₁₀H₂₀O₅Si
Boiling Pt: 190 °C
Density: 1.045 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00008593
CAS Number: 2530-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR