2-Amino-4-chloropyrimidine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-chloropyrimidin-2-amine

OR3722-5G OR3722-100G OR3722-25G
77420-716EA 23.44 USD
77420-716 77420-712 77420-714
2-Amino-4-chloropyrimidine 98%
2-Amino-4-chloropyrimidine
Formula: C₄H₄ClN₃
Melting Pt: 155…160 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00038021
CAS Number: 3993-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR