Bis(4-fluorophenyl) sulfone 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2752-100G PC2752-25G PC2752-500G
77427-808EA 74.38 USD
77427-808 77427-810 77427-812
Bis(4-fluorophenyl) sulfone 98%
Bis(4-fluorophenyl) sulphone
Formula: C₁₂H₈F₂O₂S
Melting Pt: 98…100 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000350
CAS Number: 383-29-9
EINECS: 206-847-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR