1H,1H,9H-Hexadecafluoro-1-nonanol 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1H,1H,9H-Perfluoro-1-nonanol
, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluor-1-nonanol
, 1H,1h,9h-hexadecafluorononanol
, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1-nonanol
PC4530-25G
PC4530-100G
77430-064EA
62.83
USD
77430-064
77430-066
1H,1H,9H-Hexadecafluoro-1-nonanol 97%
1H,1H,9H-Hexadecafluoro-1-nonanol
Contains upto 10% H(CF2CF2)nCHOH(CF2CF2)mH where m+n=4.
Formula:
C₉H₄F₁₆O MW: 432.1 g/mol Boiling Pt: 155…158 °C/200 mmHg Melting Pt: 53…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039629 CAS Number: 376-18-1 EINECS: 206-806-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...