5-Amino-2-fluorobenzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC50393-25G PC50393-100G
77430-916EA 38.72 USD
77430-916 77430-914
5-Amino-2-fluorobenzonitrile 95%
5-Amino-2-fluorobenzonitrile
Formula: C₇H₅FN₂
Storage Temperature: Refrigerator
MDL Number: MFCD00277872
CAS Number: 53312-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR