(Methylsulfonyl)benzene
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR2412-10G
OR2412-25G
77407-420EA
21.26
USD
77407-420
77407-426
(Methylsulfonyl)benzene
(Methylsulphonyl)benzene
Formula:
C₇H₈O₂S Melting Pt: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014741 CAS Number: 3112-85-4 EINECS: 221-476-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...