Potassium fluoride dihydrate 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC6267R-250G
PC6267R-500G
PC6267R-50G
77417-338EA
30.56
USD
77417-338
77417-340
77417-342
Potassium fluoride dihydrate 98%
Potassium fluoride dihydrate
Formula:
FK·2H₂O Boiling Pt: 156 °C Melting Pt: 41 °C Density: 2.454 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00149929 CAS Number: 13455-21-5 EINECS: 232-151-5 UN: 1812 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...