2-Fluoro-5-(trifluoromethyl)benzyl alcohol 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC4375G-100G PC4375G-25G PC4375G-5G
77429-686EA 239.92 USD
77429-686 77429-688 77429-690
2-Fluoro-5-(trifluoromethyl)benzyl alcohol 98%
2-Fluoro-5-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD00061167
CAS Number: 207974-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR