Pentafluorobenzamide 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,3,4,5,6-Pentafluorobenzamide

PC5445-25G PC5445-5G
77431-654EA 85.58 USD
77431-654 77431-656
Pentafluorobenzamide 99%
Pentafluorobenzamide
Formula: C₇H₂F₅NO
MW: 211.09 g/mol
Boiling Pt: 272
Melting Pt: 151
Storage Temperature: Ambient
MDL Number: MFCD00007971
CAS Number: 652-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR